Learn More
Gibco™ Human BMP-4 Recombinant Protein, PeproTech®
Description
120-05ET-1MG will be provided as 2 x 500 μg (120-05ET-500UG). PeproTech's E.coli-derived BMP-4 is a fully active homodimeric protein consisting of two 106 amino acid subunits, which correspond to amino acids 303-408 of the full length BMP-4 precursor. The calculated molecular weight of Recombinant Human BMP-4 (E.coli-derived) is 23.9 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.
Specifications
Specifications
| Accession Number | P12644 |
| For Use With (Application) | Control, Western Blot Control, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 652 |
| Molecular Weight (g/mol) | 23.9 kDa |
| Name | Human BMP-4 |
| Purification Method | Purified |
| Quantity | 500 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.