Learn More
Bon Opus Biosciences Recombinant Human BMP-7 (C-hFc) 50ug
Supplier: Bon Opus Biosciences BP104550UG
Recombinant Human BMP-7 (C-hFc) is produced by Human Cells. The target gene encoding Arg292-Cys430 is expressed with a C-hFc tag. Accession# P18075. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1045