Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human BMP-7 Quantikine ELISA Kit

Catalog No. SBP700
Change view
Click to view available options
Quantity:
1 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SBP700 1 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SBP700 Supplier R&D Systems™ Supplier No. SBP700
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human BMP-7 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human BMP-7 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human BMP-7 in cell culture supernates (50 ul), bone extracts (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), urine (50 ul).

Specifications

Assay Range 31.20 - 2000pg/mL
Assay Sensitivity 7.83pg/mL
Assay Solid Phase Sandwich ELISA
Conjugate HRP
Cross Reactivity Cross-reactivity observed with 1 or more available related molecules, < 50% cross-species reactivity observed with species tested
Product Type Human BMP-7 Quantikine ELISA Kit
Sample Type Bone Extracts, Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human BMP-7
For Use With (Application) ELISA
Gene ID (Entrez)
  • 655 (Human)
  • 12162 (Mouse)
  • 85272 (Rat)
Precision
  • Intra-Assay Precision (Precision within an assay) Three samples of known concentration were tested on one plate to assess intra-assay precision
  • Inter-Assay Precision (Precision between assays) Three samples of known concentration were tested in separate assays to assess inter-assay precision
Quantity 1 Pack
Sample Volume Cell Culture Supernates (50uL),Bone Extracts (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Urine (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym BMP7, BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1, OP-1 OP1, Osteogenic protein 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.