Learn More
Gibco™ Human BMP-7 Recombinant Protein, PeproTech®
Description
120-03P-1MG will be provided as 10 x 100 μg (120-03P-100UG). 120-03P-500UG will be provided as 5 x 100 μg (120-03P-100UG). Recombinant Recombinant Human BMP-7 has a calculated molecular weight of 26.4 kDa; however, due to glycosylation, the BMP-7 homodimer migrates at an apparent molecular weight of approximately 25-35 kDa by SDS-PAGE analysis under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.
Specifications
Specifications
| Accession Number | P18075 |
| For Use With (Application) | Bioactivity, Control |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 655 |
| Molecular Weight (g/mol) | 26.4 kDa |
| Name | Human BMP-7 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Source | CHO cells |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.