Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human BMP-8 ELISA Kit

Catalog No. EH48RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH48RBX5 5 x 96 Tests
EH48RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH48RBX5 Supplier Invitrogen™ Supplier No. EH48RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human BMP-8 quantitates human BMP-8 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human BMP-8.

The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. Based on its expression early in embryogenesis, the BMP encoded by this gene has a proposed role in early development. In addition, the fact that this BMP is closely related to BMP5 and BMP7 has led to speculation of possible bone inductive activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P34820, Q7Z5Y6
Assay Range 0.819-200 ng/mL
Assay Sensitivity 0.8 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias BMP; Bmp7r1; BMP8; BMP-8; BMP8A; BMP-8A; BMP8B; BMP-8B; Bone morphogenetic protein; bone morphogenetic protein 8 (osteogenic protein 2); bone morphogenetic protein 8A; Bone morphogenetic protein 8B; dJ118J21.1 (bone morphogenetic protein 8 (osteogenic protein 2)); MGC131757; OP2; OP-2; Op3; osteogenic protein 2BMP; Bmp7r1; BMP8; BMP-8; BMP8A; BMP-8A; BMP8B; BMP-8B; Bone morphogenetic protein; bone morphogenetic protein 8 (osteogenic protein 2); bone morphogenetic protein 8A; Bone morphogenetic protein 8B; dJ118J21.1 (bone morphogenetic protein 8 (osteogenic protein 2)); MGC131757; OP2; OP-2; Op3; osteogenic protein 2;
Gene ID (Entrez) 353500, 656
Gene Symbol BMP8A, BMP8B
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target BMP-8
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.