Learn More
BIOHIPPO Human BMP15 Microsample
SDP
Catalog No. 5000314545
Encompass
Human BMP15 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BMP15 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 12 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human BMP15 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human BMP15 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human BMP15 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14545 Supplier BIOHIPPO Supplier No. BHE15211833
Human BMP15 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BMP15 in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 12 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human BMP15 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human BMP15 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human BMP15 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color