Learn More
Sino Biological Anti-BMP4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10609T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human BMP4 (rh BMP4; Catalog#10609-HNAE2; P12644; Ser293-Arg408). BMP4 specific IgG was purified by Human BMP4 affinity chromatography.
Catalog No. 50-255-5249