Learn More
BIOHIPPO Human BMP6 Microsample
SDP
Catalog No. 5000314276
Encompass
Human BMP6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BMP6 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 5 6 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human BMP6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human BMP6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human BMP6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14276 Supplier BIOHIPPO Supplier No. BHE15212178
Human BMP6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BMP6 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 5 6 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human BMP6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human BMP6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human BMP6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color