Learn More
Sino Biological Anti-BMP7 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10083MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human BMP-7 (rh BMP-7; NP_001710.1; Ser 293-His 431). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-7674