Learn More
BIOHIPPO Human BMP8A Microsample
SDP
Catalog No. 5000314994
Encompass
Human BMP8A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BMP8A in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human BMP8A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human BMP8A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human BMP8A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14994 Supplier BIOHIPPO Supplier No. BHE15211355
Human BMP8A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BMP8A in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human BMP8A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human BMP8A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human BMP8A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color