Learn More
BIOHIPPO Human BMPER Microsample
SDP
Catalog No. 5000314942
Encompass
Human BMPER Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BMPER in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 054 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human BMPER Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human BMPER Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human BMPER biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14942 Supplier BIOHIPPO Supplier No. BHE15211676
Human BMPER Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BMPER in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 054 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human BMPER Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human BMPER Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human BMPER biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color