Learn More
ACROBiosystems ACROBiosystems Human BMPR-IA / ALK-3 Protein, Fc Tag
Catalog No. 502106289
Encompass_Preferred
Recombinant Protein;1MG;Human BMPR-IA, Fc Tag (AL3-H5259) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Arg 152 (Accession # P36894-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.3 kDa. The protein migrates as 56 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6289 Supplier ACROBiosystems Supplier No. AL3H52591MG
Recombinant Protein;1MG;Human BMPR-IA, Fc Tag (AL3-H5259) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Arg 152 (Accession # P36894-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.3 kDa. The protein migrates as 56 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.