Learn More
Raybiotech Inc Human BMPR-II ELISA Kit
Supplier:  Raybiotech Inc ELHBMPR21
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 2.5 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Human BMPR-2
Catalog No. 50-194-9034