Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human BMPR1A (ALK-3) Fc Chimera Recombinant Protein

Catalog No. PIRP87720
Change view
Click to view available options
Quantity:
1 mg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP87720 1 mg
PIRP87722 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIRP87720 Supplier Gibco™ Supplier No. RP87720
Add to Cart
Add to Cart
Request a Quote

Recombinant Protein

Human BMPR1A (ALK-3) Fc Chimera Recombinant Protein for Bioactivity

BMPR1A is a Type I member of the TGF beta receptor superfamily of transmembrane ser/thr kinases which phosphorylates intracellular SMADs in response to bone morphogenic proteins. These receptors are also closely related to the activin receptors, ACVR1 and ACVR2. The ligands of these receptors are members of the TGF-beta superfamily. TGF-betas and activins transduce their signals through the formation of heteromeric complexes with 2 different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kD and type II receptors of about 70-80 kD. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P36894
For Use With (Application) Bioactivity
Formulation PBS with no preservative
Gene ID (Entrez) 657
Molecular Weight (g/mol) 40.4 kDa
Name Human BMPR1A (ALK-3) Fc Chimera
Purification Method Purified
Quantity 1 mg
Source CHO cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration ≤1 EU/μg
Gene Alias 10q23del; 1110037I22Rik; activin A receptor, type II-like kinase 3; activin receptor-like kinase 3; ACVRLK3; ALK3; ALK-3; AU045487; BMP type-1A receptor; BMP-2/BMP-4 receptor; Bmpr; Bmpr1a; BMPR-1A; BMPRIA; BMPR-IA; Bone morphogenetic protein 4 receptor; bone morphogenetic protein receptor type 1A; bone morphogenetic protein receptor type IA; bone morphogenetic protein receptor type-1A; bone morphogenetic protein receptor, type 1A; bone morphogenetic protein receptor, type IA; CD292; Serine/threonine-protein kinase receptor R5; SKR5
Common Name BMPR1A
Gene Symbol BMPR1A
Biological Activity ED50 = 120 ng/mL; determined by the dose-dependent inhibition of human BMP-4-induced alkaline phosphatase production by ATDC5 cells.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence Recombinant human BMPR1A is a homodimer containing two 362 amino acid subunits with a C-terminal linker (IEGRMD) and a human IgG1 Fc-tag (AA Sequence: QNLDSMLHGT GMKSDSDQKK SENGVTLAPE DTLPFLKCYC SGHCPDDAIN NTCITNGHCF AIIEEDDQGE TTLASGCMKY EGSDFQCKDS PKAQLRRTIE CCRTNLCNQY LQPTLPPVVI GPFFDGSIRI EGRMDDKTHT CPPCPAPELL GGPSVFLFPP KPKDTLMISR TPEVTCVVVD VSHEDPEVKF NWYVDGVEVH NAKTKPREEQ YNSTYRVVSV LTVLHQDWLN GKEYKCKVSN KALPAPIEKT ISKAKGQPRE PQVYTLPPSR DELTKNQVSL TCLVKGFYPS DIAVEWESNG QPENNYKTTP PVLDSDGSFF LYSKLTVDKS RWQQGNVFSC SVMHEALHNH YTQKSLSLSP GK)
Content And Storage -20°C
Activity ED50 ≤ 120 ng/mL; determined by the dose-dependent inhibition of human BMP-4-induced alkaline phosphatase production by ATDC5 cells.
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade >95% by SDS-Page
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.