Learn More
Sino Biological Recombinant Human BMPR1A/ALK-3 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 10446H02H100
Encompass_Preferred
A DNA sequence encoding the human BMPR1A (NP_004320.2) (Met1-Arg152) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-5301