Learn More
Sino Biological Anti-BMPRIB Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10460RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ALK-6 / BMPR1B (rh BMPR1B; Catalog#10460-H20B; NP_001194.1; Arg149-Leu502). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8923