Learn More
Sino Biological Recombinant Human BMPR2 Protein (His Tag), HPLC-verified 1 mg
Supplier:  Sino Biological 10551H08H1
Encompass_Preferred
A DNA sequence encoding the human BMPR-II (NP_001195.2) extracellular domain (Met 1-Ile 151) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-7030