Learn More
Sino Biological Anti-BMPR2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10551RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human BMPR2 (rh BMPR2; Catalog#10551-H08H; NP_001195.2; Met 1-Ile 151). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2019