Learn More
Sino Biological Human BMPR2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10551T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human BMPR2 (rh BMPR2; Catalog#10551-H08H; NP_001195.2; Met1-Ile151). BMPR2 specific IgG was purified by Human BMPR2 affinity chromatography.
Catalog No. 50-237-2909