Learn More
Lonza Walkersville Unprocessed Human Bone Marrow, Fresh, 10 mL
Supplier:  Lonza Walkersville 1M105
Encompass
We Find the Donor. You Find the Cure.At Lonza, we are committed to handling the logistics so you can focus on finding the cure. Providing the research community with fresh unprocessed bone marrow while maintaining the well-being of our donors is at the forefront of Lonza's approved bone marrow donor program. We have been delivering the fresh unprocessed bone marrow you need for over 20 years so you can focus on the important work behind finding the cure.More Cells:All research-use, fresh unprocessed bone marrow samples contain greater than 15 million nucleated viable cells per mL, giving you more cells for your money. A total of 100 mL per donor can be ordered in 10 or 25 mL quantities. Up to 100 mL can be ordered from each donor.Benefits:Test negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsCertificate of Analysis (CoA) is provided for each cell lot purchased
Catalog No. NC1342182