Learn More
Lonza Walkersville Human Bone Marrow Fresh 25mL
Catalog No. 5000496298
Encompass
All research-use fresh unprocessed bone marrow samples contain greater than 15 million nucleated viable cells per mL giving you more cells for your money A total of 100 mL per donor can be ordered in 10 or 25 mL quantities Up to 100 mL can be ordered from each donor
Catalog No. 50-004-96298 Supplier Lonza Walkersville Supplier No. 1M125
All research-use fresh unprocessed bone marrow samples contain greater than 15 million nucleated viable cells per mL giving you more cells for your money A total of 100 mL per donor can be ordered in 10 or 25 mL quantities Up to 100 mL can be ordered from each donor