Learn More
STEMCELL Technologies Human Bone Marrow Mononuclear Cells, Frozen, Size: 5 x 10^6 cells
SDP
Catalog No. 501975767
Encompass
Primary bone marrow mononuclear cells (a.k.a. BM MNCs or BMNCs) are a heterogeneous population that includes hematopoietic lineage cells such as lymphocytes, monocytes, stem cells, and progenitor cells as well as mesenchymal stromal cells. Mononuclear cells are isolated from human adult bone marrow using density gradient separation. Heparin is added to the bone marrow upon collection as an anticoagulant.Cells were obtained using Institutional Review Board (IRB) approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.
Catalog No. 50-197-5767 Supplier STEMCELL Technologies Supplier No. 70001.1
Primary bone marrow mononuclear cells (a.k.a. BM MNCs or BMNCs) are a heterogeneous population that includes hematopoietic lineage cells such as lymphocytes, monocytes, stem cells, and progenitor cells as well as mesenchymal stromal cells. Mononuclear cells are isolated from human adult bone marrow using density gradient separation. Heparin is added to the bone marrow upon collection as an anticoagulant.Cells were obtained using Institutional Review Board (IRB) approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.