Learn More
Biomatik Corporation Human Bone Morphogenetic Protein 8A (BMP8A) ELISA Kit
Supplier:  Biomatik Corporation EKU08496
Encompass_Preferred
Human Bone Morphogenetic Protein 8A (BMP8A) ELISA Kit for detecting BMP8A in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.62-1000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q7Z5Y6. Gene ID: 353500. Target Synonyms: BMP8-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5373