Learn More
Biomatik Corporation Human Bone Morphogenetic Protein 5 (BMP5) ELISA Kit
Supplier:  Biomatik Corporation EKU08495
Encompass_Preferred
Human Bone Morphogenetic Protein 5 (BMP5) ELISA Kit for detecting BMP5 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.62-1000pg/ml. Sensitivity: 5.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P22003. Gene ID: 653. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5725