Learn More
Biomatik Corporation Human Bone Morphogenetic Protein 9 (BMP9) CLIA Kit
Supplier:  Biomatik Corporation EKU09694
Encompass_Preferred
Human Bone Morphogenetic Protein 9 (BMP9) CLIA Kit for detecting BMP9 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 68.6-50,000pg/ml. Sensitivity: 27.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9UK05. Gene ID: 2658. Target Synonyms: GDF2; Growth Differentiation Factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7279