Learn More
Biomatik Corporation Human Bone Morphogenetic Protein 8B (BMP8B) ELISA Kit
Supplier:  Biomatik Corporation EKU08497
Encompass_Preferred
Human Bone Morphogenetic Protein 8B (BMP8B) ELISA Kit for detecting BMP8B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P34820. Gene ID: 656. Target Synonyms: BMP8-B; OP2; Osteogenic Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8660