Learn More
Biomatik Corporation Human Bone Morphogenetic Protein 2 (BMP2) CLIA Kit
Supplier:  Biomatik Corporation EKU09527
Encompass_Preferred
Human Bone Morphogenetic Protein 2 (BMP2) CLIA Kit for detecting BMP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.55pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P12643. Gene ID: 650. Target Synonyms: BMP2A; BMP-2A; Hemochromatosis Modifier. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5116