Learn More
Sino Biological Anti-BPI Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13907RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human BPI (rh BPI; Catalog#13907-H08H; P17213; Met1-Lys487). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4164