Learn More
Sino Biological Human BPIFB1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 13275T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human BPIFB1 / LPLUNC1 (rh BPIFB1 / LPLUNC1; Catalog#13275-H08H; AAH08429.1; Met1-Gln484). BPIFB1 / LPLUNC1 specific IgG was purified by Human BPIFB1 / LPLUNC1 affinity chromatography.
Catalog No. 50-237-2551