Learn More
Biomatik Corporation Human Brain Natriuretic Peptide (BNP) CLIA Kit
Supplier:  Biomatik Corporation EKU09466
Encompass_Preferred
Human Brain Natriuretic Peptide (BNP) CLIA Kit for detecting BNP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.86-5,000pg/ml. Sensitivity: 2.56pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P16860. Gene ID: 4879. Target Synonyms: GC-B; B-Type Natriuretic Peptide; Ventricular Natriuretic Peptide; Gamma-brain natriuretic peptide; Brain natriuretic peptide 32. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6534