Learn More
Biomatik Corporation Human Brain Specific Angiogenesis Inhibitor 3 (BAI3) ELISA Kit
Supplier:  Biomatik Corporation EKU02799
Encompass_Preferred
Human Brain Specific Angiogenesis Inhibitor 3 (BAI3) ELISA Kit for detecting BAI3 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60242. Gene ID: 577. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9094