Learn More
Biomatik Corporation Human Breast Cancer Anti-Estrogen Resistance 1 (BCAR1) ELISA Kit
Supplier:  Biomatik Corporation EKU02805
Encompass_Preferred
Human Breast Cancer Anti-Estrogen Resistance 1 (BCAR1) ELISA Kit for detecting BCAR1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.12-5000pg/ml. Sensitivity: 27pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P56945. Gene ID: 9564. Target Synonyms: CAS; CRKAS; P130Cas; CASS1; Crk-Associated Substrate; Cas Scaffolding Protein Family Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9093