Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Brevican ELISA Kit

Catalog No. EH52RBX5
Encompass_Preferred
Change view
Click to view available options
5 x 96 Tests; HRP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
EH52RBX5 5 x 96 Tests; HRP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH52RBX5 Supplier Invitrogen™ Supplier No. EH52RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human Brevican quantitates human Brevican in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Brevican.

Brevican is the most abundant chondroitin sulfate proteoglycan in the extracellular matrix of the adult brain. It is a member of the lectican family of aggregating extracellular matrix proteoglycans that bear chondroitin sulfate (CS) chains. It is highly expressed in the central nervous system and is thought to stabilize synapses and inhibit neural plasticity. Brevican is secreted from astrocytes and neurons as a 145 kD core protein that bears up to three, covalently-linked, CS chains. It is also is secreted as a 145 kD core protein without CS chains. When cleaved by extracellular glutamyl endopeptidases, the ADAMTSs, a 55 kD N-terminal fragment is formed that contains the unique C terminal epitope EAMESE.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96GW7
Assay Range 0.041-10 ng/mL
Assay Sensitivity 0.041 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias ALPBRE; Bcan; Behab; Brain-enriched hyaluronan-binding protein; brevican; brevican (brain specific proteoglycan in the aggrecan family); brevican core protein; Chondroitin sulfate proteoglycan 7; chondroitin sulfate proteoglycan BEHAB; CSPG7; UNQ2525/PRO6018ALPBRE; Bcan; Behab; Brain-enriched hyaluronan-binding protein; brevican; brevican (brain specific proteoglycan in the aggrecan family); brevican core protein; Chondroitin sulfate proteoglycan 7; chondroitin sulfate proteoglycan BEHAB; CSPG7; UNQ2525/PRO6018;
Gene ID (Entrez) 63827
Gene Symbol Bcan
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Brevican
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.