Learn More
Biomatik Corporation Human Bridging Integrator 1 (BIN1) ELISA Kit
Supplier:  Biomatik Corporation EKU02811
Encompass_Preferred
Human Bridging Integrator 1 (BIN1) ELISA Kit for detecting BIN1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00499. Gene ID: 274. Target Synonyms: AMPH2; AMPHL; SH3P9; Amphiphysin II; Myc Box-Dependent-Interacting Protein 1; Amphiphysin-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4565