Learn More
Biomatik Corporation Human Bridging Integrator 2 (BIN2) ELISA Kit
Supplier:  Biomatik Corporation EKU02812
Encompass_Preferred
Human Bridging Integrator 2 (BIN2) ELISA Kit for detecting BIN2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UBW5. Gene ID: 51411. Target Synonyms: BRAP-1; Breast cancer-associated protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9092