Learn More
Biomatik Corporation Human Bromodomain Containing Protein 9 (BRD9) ELISA Kit
Supplier:  Biomatik Corporation EKU11851
Encompass_Preferred
Human Bromodomain Containing Protein 9 (BRD9) ELISA Kit for detecting BRD9 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H8M2. Gene ID: 65980. Target Synonyms: Rhabdomyosarcoma antigen MU-RMS-40.8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8070