Learn More
Biomatik Corporation Human Bromodomain Containing Protein 2 (BRD2) ELISA Kit
Supplier:  Biomatik Corporation EKU12438
Encompass_Preferred
Human Bromodomain Containing Protein 2 (BRD2) ELISA Kit for detecting BRD2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P25440. Target Synonyms: FSRG1; NAT; RING3; RNF3; Really interesting new gene 3 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7965