Learn More
Abcam BSA ELISA Kit, 96TEST
Supplier:  Abcam AB28524596TEST
Encompass_Preferred
BSA ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of BSA in Human in Plasma, Tissue Homogenate, Cell culture supernatant, Serum, Other biological fluids samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5496