Learn More
Sino Biological Anti-CD147/Basigin/EMMPRIN Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10186MM0520
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD147. (rh CD147; Catalog#10186-H08H; NP_940991.1; Met 1-His 205). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-9374