Learn More
Sino Biological Human BST2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 13370T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human BST2 (rh BST2; Catalog#13370-H07H; Q10589; Asn49-Ser160). BST2 specific IgG was purified by Human BST2 affinity chromatography.
Catalog No. 50-237-2615