Learn More
Sino Biological Anti-BTK Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10578T32100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human BTK (rh BTK; Catalog#10578-H08B; NP_000052.1; Met 1-Ser 659). BTK specific IgG was purified by Human BTK affinity chromatography.
Catalog No. 50-255-5255