Learn More
Sino Biological Recombinant Human BTLA Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 30120H02H100
Encompass_Preferred
A DNA sequence encoding the Human BTLA isoform1 (NP_861445.4) (Mer1-Ser150) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-240-9056