Learn More
Sino Biological Recombinant Human BTLA Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11895H08H1
Encompass_Preferred
A DNA sequence encoding the Human BTLA (NP_001078826.1) (Met1-Thr134) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4493