Learn More
Sino Biological Recombinant Human BTLA Protein (hFc & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 11895H41HB20
Encompass_Preferred
A DNA sequence encoding the human BTLA (NP_001078826.1) (Met1-Thr134) was expressed with a C-terminal Fc region of human IgG1 tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4491