Learn More
Sino Biological Recombinant Human BTLA Protein (His & AVI Tag), Biotinylated, isoform 1,HPLC-verified
Supplier:  Sino Biological 30120H49HB20
Encompass_Preferred
A DNA sequence encoding the human BTLA (NM_181780.4) (Mer1-Ser150) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-0953