Learn More
Sino Biological Human BTN1A1 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 30072H02H1
Encompass_Preferred
A DNA sequence encoding the human BTN1A1 (Q4VAN1-1) (Met1-Arg242) was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-0960