Learn More
Sino Biological Recombinant Human CD277/BTN3A1 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 15973H08HB100
Encompass_Preferred
A DNA sequence encoding the human BTN3A1 (NP_919423.1) (Met1-Gly254) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4077