Learn More
Sino Biological Anti-BTN3A3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13432RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human BTN3A3 (rh BTN3A3; Catalog#13432-H08H; O00478-1; Met1-Trp248). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4243