Learn More
Abcam Human c-Kit ELISA Kit (CD117), 2 x 96 Tests
Supplier:  Abcam AB459242X96TESTS
Encompass_Preferred
Human c-Kit ELISA Kit (CD117) is a Sandwich (quantitative) ELISA kit for the measurement of Human c-Kit (CD117) in Human in Plasma, Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2316554