Learn More
Biomatik Corporation Human C Reactive Protein (CRP) CLIA Kit
Supplier:  Biomatik Corporation EKU08918
Encompass_Preferred
Human C Reactive Protein (CRP) CLIA Kit for detecting CRP in serum, plasma, tissue homogenates, cell lysates, urine, cerebrospinal fluid, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P02741. Gene ID: 1401. Target Synonyms: C-RP; PTX1; Pentraxin-Related; Pentraxin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5867